siRNA Details
| Gene Symbol: CCDC144A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014695.2 | NP_055510.1 | 14 | 3784 | ||
| NR_130142.1 | 13 | 3609 | ||||
| XM_017025429.1 | XP_016880918.1 | 15 | 3939 | |||
| XM_017025430.1 | XP_016880919.1 | 14 | 3874 | |||
| GenBank mRNA | AB011137.2 | - | 5 | 2186 | ||
| AK125971.1 | - | 11 | 2295 | |||
| BC133019.1 | - | 14 | 3784 | 40147041 | ||
| BC144224.1 | - | 13 | 3609 | 9052744 | ||
| BC144225.1 | - | 14 | 3784 | 9052745 | ||

CCDC144A
Genomic Map