siRNA Details
| Gene Symbol: FAM153A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173663.3 | NP_775934.3 | 20 | 1681 | ||
| Gene Symbol: FAM153B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265615.1 | NP_001252544.1 | 20 | 1681 | ||

FAM153A
Genomic Map