siRNA Details
| Gene Symbol: VWA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136043.2 | NP_001129515.1 | 8 | 1081 | ||
| NM_001207058.1 | NP_001193987.1 | 8 | 1057 | |||
| NM_001207059.1 | NP_001193988.1 | 7 | 940 | |||
| NR_045104.1 | 8 | 1224 | ||||
| NR_045105.2 | 8 | 1478 | ||||
| NR_045106.2 | 8 | 1385 | ||||
| NR_045901.1 | 7 | 1116 | ||||
| XM_006720708.3 | XP_006720771.1 | 8 | 1160 | |||
| XM_006720709.3 | XP_006720772.1 | 8 | 1225 | |||
| XM_006720711.2 | XP_006720774.1 | 7 | 1084 | |||
| XM_011522101.2 | XP_011520403.1 | 8 | 906 | |||
| XM_017022644.1 | XP_016878133.1 | 8 | 1305 | |||
| GenBank mRNA | AK027475.1 | - | 8 | 1066 | ||
| AK293207.1 | - | 7 | 893 | |||
| AK296134.1 | - | 8 | 1451 | |||
| AK301033.1 | - | 8 | 923 | |||
| AK301222.1 | - | 7 | 1069 | |||
| AK301742.1 | - | 8 | 899 | |||
| AL136662.1 | - | 8 | 1306 | |||
| BC007991.2 | - | 8 | 1051 | 3140504 | ||

VWA9
Genomic Map