siRNA Details
| Gene Symbol: C7orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100159.2 | NP_001093629.1 | 5 | 601 | ||
| NM_001267865.1 | NP_001254794.1 | 4 | 445 | |||
| NM_001267866.1 | NP_001254795.1 | 4 | 445 | |||
| XM_006715649.3 | XP_006715712.1 | 4 | 461 | |||
| XM_006715650.2 | XP_006715713.1 | 4 | 428 | |||
| XM_011515121.1 | XP_011513423.1 | 5 | 505 | |||
| XM_011515122.2 | XP_011513424.1 | 4 | 461 | |||
| XM_011515123.2 | XP_011513425.1 | 4 | 461 | |||

C7orf57
Genomic Map