siRNA Details
| Gene Symbol: GCNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080615.1 | NP_542182.1 | 5 | 1274 | ||
| GenBank mRNA | AK131203.1 | - | 5 | 1274 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCNT7| Gene Symbol: GCNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080615.1 | NP_542182.1 | 5 | 1274 | ||
| GenBank mRNA | AK131203.1 | - | 5 | 1274 | ||