siRNA Details
| Gene Symbol: ZNF626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076675.2 | NP_001070143.1 | 3 | 343 | ||
| NM_145297.3 | NP_660340.1 | 3 | 343 | |||
| GenBank mRNA | BC007116.1 | - | 3 | 322 | ||
| BC107803.1 | - | 3 | 347 | |||
| BC150651.1 | - | 3 | 254 | 9057022 | ||
| CR749233.1 | - | 3 | 295 | |||

ZNF626
Genomic Map