siRNA Details
| Gene Symbol: NBPF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170755.2 | NP_001164226.1 | 22 | 3229 | ||
| NM_173638.4 | NP_775909.2 | 19 | 2699 | |||
| XM_017001077.1 | XP_016856566.1 | 17 | 2347 | |||
| GenBank mRNA | AK290142.1 | - | 11 | 1401 | ||
| AK290302.1 | - | 16 | 2158 | |||
| AK299360.1 | - | 13 | 1739 | |||
| BC023087.1 | - | 19 | 2556 | 3918932 | ||

NBPF15
Genomic Map