siRNA Details
| Gene Symbol: OST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134693.1 | NP_001128165.1 | 2 | 186 | ||
| GenBank mRNA | AL049447.1 | - | 2 | 112 | ||
| BC105283.1 | - | 2 | 144 | |||
| BF697310.1 | - | 2 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OST4| Gene Symbol: OST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134693.1 | NP_001128165.1 | 2 | 186 | ||
| GenBank mRNA | AL049447.1 | - | 2 | 112 | ||
| BC105283.1 | - | 2 | 144 | |||
| BF697310.1 | - | 2 | 186 | |||