siRNA Details
| Gene Symbol: RBM27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018989.1 | NP_061862.1 | 12 | 1939 | ||
| XM_005268466.1 | XP_005268523.1 | 12 | 1992 | |||
| XM_005268467.1 | XP_005268524.1 | 11 | 1827 | |||
| XM_005268468.1 | XP_005268525.1 | 11 | 1824 | |||
| XM_006714787.1 | XP_006714850.1 | 13 | 1956 | |||
| XM_011537661.1 | XP_011535963.1 | 12 | 1992 | |||
| XM_011537662.1 | XP_011535964.1 | 12 | 1989 | |||
| XM_017009587.1 | XP_016865076.1 | 13 | 1953 | |||
| GenBank mRNA | AB037732.1 | - | 8 | 1262 | ||
| AK128738.1 | - | 11 | 1824 | |||
| BX647384.1 | - | 12 | 1939 | |||

RBM27
Genomic Map