siRNA Details
| Gene Symbol: ARMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105247.1 | NP_001098717.1 | 6 | 3593 | ||
| NM_001288767.1 | NP_001275696.1 | 8 | 3473 | |||
| NM_001301820.1 | NP_001288749.1 | 7 | 3608 | |||
| NM_024742.2 | NP_079018.1 | 4 | 4651 | |||
| XM_006721091.2 | XP_006721154.1 | 7 | 3744 | |||
| GenBank mRNA | AK023125.1 | - | 5 | 2740 | ||
| AK024430.1 | - | 4 | 4144 | |||
| AK297115.1 | - | 7 | 2052 | |||
| AY217348.1 | - | 4 | 4651 | |||
| BC014945.1 | - | 7 | 2998 | 4860377 | ||

ARMC5
Genomic Map