siRNA Details
| Gene Symbol: SERPINB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002639.4 | NP_002630.2 | 7 | 1201 | ||
| XM_006722483.2 | XP_006722546.1 | 4 | 709 | |||
| GenBank mRNA | AK301667.1 | - | 6 | 965 | ||
| AK312765.1 | - | 7 | 1179 | |||
| BM955647.1 | - | 2 | 429 | |||
| BX640597.1 | - | 2 | 2597 | |||
| U04313.1 | - | 7 | 1134 | |||

SERPINB5
Genomic Map