siRNA Details
| Gene Symbol: GBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098834.2 | NP_001092304.1 | 1 | 510 | ||
| XM_017011964.1 | XP_016867453.1 | 1 | 153 | |||
| XM_017011965.1 | XP_016867454.1 | 1 | 153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GBX1| Gene Symbol: GBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098834.2 | NP_001092304.1 | 1 | 510 | ||
| XM_017011964.1 | XP_016867453.1 | 1 | 153 | |||
| XM_017011965.1 | XP_016867454.1 | 1 | 153 | |||