siRNA Details
| Gene Symbol: VWA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009814.1 | NP_001009814.1 | 12 | 1459 | ||
| NM_015058.1 | NP_055873.1 | 12 | 1459 | |||
| XM_006719791.3 | XP_006719854.1 | 4 | 415 | |||
| XM_011535007.2 | XP_011533309.1 | 12 | 1485 | |||
| XM_017020469.1 | XP_016875958.1 | 14 | 1518 | |||
| XM_017020470.1 | XP_016875959.1 | 12 | 1485 | |||
| XM_017020471.1 | XP_016875960.1 | 12 | 1485 | |||
| XM_017020474.1 | XP_016875963.1 | 12 | 1485 | |||
| GenBank mRNA | AK095155.1 | - | 12 | 1333 | ||
| AL834181.1 | - | 5 | 508 | |||
| BC053674.1 | - | 12 | 1459 | 6171968 | ||
| BC131802.1 | - | 13 | 1292 | |||

VWA8
Genomic Map