siRNA Details
| Gene Symbol: SSX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021014.3 | NP_066294.1 | 6 | 503 | ||
| XM_011543885.2 | XP_011542187.1 | 6 | 456 | |||
| GenBank mRNA | AK310513.1 | - | 6 | 456 | ||
| BC005904.1 | - | 6 | 457 | 4064072 | ||
| BC103862.1 | - | 6 | 502 | 40003336 | ||
| S82471.1 | - | 5 | 363 | |||
| U90840.1 | - | 6 | 416 | |||

SSX3
Genomic Map