siRNA Details
| Gene Symbol: HIGD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109619.1 | NP_001103089.1 | 2 | 131 | ||
| XM_011538649.2 | XP_011536951.1 | 4 | 278 | |||
| XM_017019783.1 | XP_016875272.1 | 5 | 1706 | |||
| XM_017019784.1 | XP_016875273.1 | 5 | 1706 | |||
| XM_017019785.1 | XP_016875274.1 | 3 | 210 | |||
| GenBank mRNA | DW009649.1 | - | 2 | 68 | ||

HIGD1C
Genomic Map