siRNA Details
Gene Symbol: PRR32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122716.1 | NP_001116188.1 | 2 | 544 | ||
GenBank mRNA | BG200532.1 | - | 1 | 444 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRR32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122716.1 | NP_001116188.1 | 2 | 544 | ||
GenBank mRNA | BG200532.1 | - | 1 | 444 |