siRNA Details
| Gene Symbol: ARSI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012301.2 | NP_001012301.1 | 2 | 1220 | ||
| GenBank mRNA | DA264664.1 | - | 2 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARSI| Gene Symbol: ARSI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012301.2 | NP_001012301.1 | 2 | 1220 | ||
| GenBank mRNA | DA264664.1 | - | 2 | 350 | ||