siRNA Details
| Gene Symbol: C5orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001089584.2 | NP_001083053.1 | 3 | 1210 | ||
| XM_011513963.2 | XP_011512265.1 | 3 | 761 | |||
| GenBank mRNA | BC053548.1 | - | 3 | 863 | ||
| BC062569.1 | - | 3 | 1210 | |||
| BC104204.2 | - | 3 | 865 | |||
| BC104205.2 | - | 3 | 871 | 40031408 | ||
| BC115361.1 | - | 3 | 871 | 40031409 | ||

C5orf49
Genomic Map