siRNA Details
| Gene Symbol: HIST1H2AI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003509.2 | NP_003500.1 | 1 | 415 | ||
| GenBank mRNA | BC112254.1 | - | 1 | 493 | 8327722 | |
| BC112256.1 | - | 1 | 493 | 8327724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AI| Gene Symbol: HIST1H2AI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003509.2 | NP_003500.1 | 1 | 415 | ||
| GenBank mRNA | BC112254.1 | - | 1 | 493 | 8327722 | |
| BC112256.1 | - | 1 | 493 | 8327724 | ||