siRNA Details
| Gene Symbol: TRIM35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304495.1 | NP_001291424.1 | 2 | 560 | ||
| NM_171982.4 | NP_741983.2 | 3 | 656 | |||
| XM_005273452.3 | XP_005273509.1 | 3 | 651 | |||
| XM_011544453.1 | XP_011542755.1 | 4 | 684 | |||
| GenBank mRNA | AB029021.1 | - | 3 | 606 | ||
| AF492463.1 | - | 3 | 654 | |||
| AK308236.1 | - | 2 | 480 | |||
| BC018337.1 | - | 4 | 766 | |||
| BC069226.1 | - | 3 | 598 | 6428667 | ||

TRIM35
Genomic Map