siRNA Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021732.2 | NP_068378.2 | 2 | 641 | ||
| XM_017016494.1 | XP_016871983.1 | 2 | 801 | |||
| GenBank mRNA | AF131791.1 | - | 2 | 641 | ||
| AF241786.1 | - | 2 | 643 | |||
| AK024533.1 | - | 2 | 613 | |||
| AK222493.1 | - | 2 | 614 | |||

AVPI1
Genomic Map