siRNA Details
| Gene Symbol: ZNF257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316996.1 | NP_001303925.1 | 5 | 873 | ||
| NM_001316997.1 | NP_001303926.1 | 5 | 817 | |||
| NM_001316998.1 | NP_001303927.1 | 2 | 529 | |||
| NM_033468.3 | NP_258429.2 | 4 | 752 | |||
| GenBank mRNA | AK056694.1 | - | 4 | 637 | ||
| BC036446.1 | - | 5 | 867 | 4831021 | ||
| BU155753.1 | - | 5 | 729 | |||
| BX100770.1 | - | 2 | 529 | |||
| DB134269.1 | - | 2 | 494 | |||

ZNF257
Genomic Map