siRNA Details
| Gene Symbol: LKAAEAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007125.1 | NP_001007126.1 | 2 | 852 | ||
| XM_005260200.3 | XP_005260257.3 | 4 | 694 | |||
| GenBank mRNA | BC036837.1 | - | 2 | 852 | 5173095 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LKAAEAR1| Gene Symbol: LKAAEAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007125.1 | NP_001007126.1 | 2 | 852 | ||
| XM_005260200.3 | XP_005260257.3 | 4 | 694 | |||
| GenBank mRNA | BC036837.1 | - | 2 | 852 | 5173095 | |