siRNA Details
| Gene Symbol: SPINT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006652.1 | NP_006643.1 | 1 | 83 | ||
| GenBank mRNA | AY372172.2 | - | 1 | 83 | ||
| BX088813.1 | - | 1 | 33 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPINT3| Gene Symbol: SPINT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006652.1 | NP_006643.1 | 1 | 83 | ||
| GenBank mRNA | AY372172.2 | - | 1 | 83 | ||
| BX088813.1 | - | 1 | 33 | |||