siRNA Details
| Gene Symbol: PNMAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 534 | ||
| NM_018215.3 | NP_060685.2 | 2 | 534 | |||
| XM_011527067.2 | XP_011525369.1 | 2 | 548 | |||
| GenBank mRNA | AK096326.1 | - | 1 | 159 | ||
| AK290218.1 | - | 2 | 482 | |||
| AL834344.1 | - | 2 | 378 | |||
| BC032508.1 | - | 2 | 512 | 5246377 | ||
| BC051688.1 | - | 2 | 491 | 5295669 | ||
| DA219062.1 | - | 2 | 509 | |||
| DA325514.1 | - | 2 | 534 | |||

PNMAL1
Genomic Map