siRNA Details
| Gene Symbol: KHDC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126063.2 | NP_001119535.1 | 1 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KHDC1L| Gene Symbol: KHDC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126063.2 | NP_001119535.1 | 1 | 94 | ||