siRNA Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040152.1 | NP_001035242.1 | 2 | 2467 | ||
| NM_001172437.2 | NP_001165908.1 | 2 | 2478 | |||
| NM_001172438.2 | NP_001165909.1 | 2 | 2478 | |||
| NM_001184961.1 | NP_001171890.1 | 2 | 2467 | |||
| NM_001184962.1 | NP_001171891.1 | 2 | 2467 | |||
| NM_015068.3 | NP_055883.2 | 2 | 2467 | |||
| GenBank mRNA | AB028974.1 | - | 1 | 2040 | ||
| AB049150.1 | - | 2 | 2246 | |||
| AB049834.1 | - | 1 | 2105 | |||
| AF216076.1 | - | 1 | 1998 | |||
| AK299837.1 | - | 2433 | ||||

PEG10
Genomic Map