siRNA Details
| Gene Symbol: CCDC152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134848.1 | NP_001128320.1 | 3 | 185 | ||
| GenBank mRNA | AK127368.1 | - | 3 | 175 | ||
| AK303469.1 | - | 3 | 185 | |||
| AY947526.1 | - | 2 | 116 | |||
| BC015985.1 | - | 3 | 178 | |||
| BC039102.1 | - | 3 | 157 | |||

CCDC152
Genomic Map