siRNA Details
| Gene Symbol: C1orf68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024679.2 | NP_001019850.1 | 1 | 232 | ||
| GenBank mRNA | CU446285.1 | - | 1 | 72 | ||
| CU457096.1 | - | 1 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf68| Gene Symbol: C1orf68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024679.2 | NP_001019850.1 | 1 | 232 | ||
| GenBank mRNA | CU446285.1 | - | 1 | 72 | ||
| CU457096.1 | - | 1 | 195 | |||