siRNA Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1589 | ||
| XM_011526858.2 | XP_011525160.1 | 2 | 1945 | |||
| GenBank mRNA | BC035657.1 | - | 2 | 1575 | 5588752 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FFAR3| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1589 | ||
| XM_011526858.2 | XP_011525160.1 | 2 | 1945 | |||
| GenBank mRNA | BC035657.1 | - | 2 | 1575 | 5588752 | |