siRNA Details
| Gene Symbol: GLRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024452.2 | NP_001019623.2 | 2 | 612 | ||
| NM_001172285.1 | NP_001165756.1 | 2 | 612 | |||
| GenBank mRNA | AK309010.1 | - | 2 | 612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLRA4| Gene Symbol: GLRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024452.2 | NP_001019623.2 | 2 | 612 | ||
| NM_001172285.1 | NP_001165756.1 | 2 | 612 | |||
| GenBank mRNA | AK309010.1 | - | 2 | 612 | ||