siRNA Details
| Gene Symbol: GTF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002097.2 | NP_002088.2 | 7 | 876 | ||
| GenBank mRNA | AK057993.1 | - | 7 | 1412 | ||
| BC143711.1 | - | 6 | 868 | |||
| D32257.1 | - | 7 | 875 | |||
| U14134.1 | - | 6 | 601 | |||
| U20272.1 | - | 7 | 676 | |||

GTF3A
Genomic Map