siRNA Details
| Gene Symbol: KRTAP10-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198689.2 | NP_941962.1 | 1 | 1549 | ||
| GenBank mRNA | AJ566385.1 | - | 1 | 575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP10-7| Gene Symbol: KRTAP10-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198689.2 | NP_941962.1 | 1 | 1549 | ||
| GenBank mRNA | AJ566385.1 | - | 1 | 575 | ||