siRNA Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198989.3 | NP_945340.2 | 2 | 1215 | ||
| GenBank mRNA | AK126830.1 | - | 2 | 1215 | ||
| BC104892.1 | - | 2 | 1025 | 8143895 | ||
| BC112009.1 | - | 2 | 1025 | 8327477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLEU7| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198989.3 | NP_945340.2 | 2 | 1215 | ||
| GenBank mRNA | AK126830.1 | - | 2 | 1215 | ||
| BC104892.1 | - | 2 | 1025 | 8143895 | ||
| BC112009.1 | - | 2 | 1025 | 8327477 | ||