siRNA Details
| Gene Symbol: PWWP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098637.1 | NP_001092107.1 | 3 | 2211 | ||
| NM_138499.3 | NP_612508.3 | 3 | 2514 | |||
| XM_011539387.2 | XP_011537689.1 | 4 | 2331 | |||
| GenBank mRNA | AK096940.1 | - | 1 | 2746 | ||
| AK128663.1 | - | 2 | 2350 | |||
| BC011630.2 | - | 2 | 1986 | |||
| BC068574.1 | - | 3 | 2512 | 30344364 | ||

PWWP2B
Genomic Map