siRNA Details
| Gene Symbol: ZSCAN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 925 | ||
| XM_005248950.2 | XP_005249007.1 | 4 | 923 | |||
| XM_011514406.2 | XP_011512708.1 | 4 | 923 | |||
| XM_011514408.2 | XP_011512710.1 | 5 | 1110 | |||
| XM_011514410.2 | XP_011512712.1 | 4 | 923 | |||
| XM_017010479.1 | XP_016865968.1 | 4 | 4139 | |||
| GenBank mRNA | AK092117.1 | - | 3 | 755 | ||
| AK309235.1 | - | 2 | 242 | |||
| AL833347.1 | - | 4 | 890 | |||
| BC101557.1 | - | 3 | 827 | 8069063 | ||
| BC101559.1 | - | 3 | 827 | 8069065 | ||

ZSCAN23
Genomic Map