siRNA Details
| Gene Symbol: XAGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130777.2 | NP_570133.1 | 5 | 574 | ||
| XM_011530836.2 | XP_011529138.1 | 5 | 602 | |||
| GenBank mRNA | AJ318880.1 | - | 5 | 549 | ||
| BC009232.2 | - | 5 | 554 | 3960352 | ||
| BC066311.1 | - | 5 | 556 | 30330532 | ||

XAGE2
Genomic Map