siRNA Details
| Gene Symbol: TRAF3IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139490.1 | NP_001132962.1 | 3 | 421 | ||
| NM_015650.3 | NP_056465.2 | 3 | 421 | |||
| XM_006712414.2 | XP_006712477.1 | 3 | 476 | |||
| XM_011510944.2 | XP_011509246.1 | 3 | 476 | |||
| XM_011510945.2 | XP_011509247.1 | 3 | 476 | |||
| XM_011510946.2 | XP_011509248.1 | 3 | 476 | |||
| XM_011510947.2 | XP_011509249.1 | 3 | 463 | |||
| XM_011510948.2 | XP_011509250.1 | 3 | 476 | |||
| XM_017003789.1 | XP_016859278.1 | 3 | 476 | |||
| GenBank mRNA | AF230877.1 | - | 3 | 302 | ||
| AF242456.1 | - | 3 | 520 | |||
| AL080153.1 | - | 2 | 159 | |||
| BC059174.1 | - | 3 | 421 | 6191726 | ||

TRAF3IP1
Genomic Map