siRNA Details
| Gene Symbol: C19orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317801.1 | NP_001304730.1 | 4 | 387 | ||
| NM_033520.2 | NP_277055.1 | 4 | 328 | |||
| GenBank mRNA | AB038317.1 | - | 4 | 325 | ||
| AB038318.1 | - | 4 | 384 | |||
| AF213678.1 | - | 4 | 328 | |||
| BC008888.2 | - | 4 | 285 | |||
| BC014264.2 | - | 4 | 285 | 4550805 | ||
| BC060319.1 | - | 4 | 339 | 3901226 | ||
| BX648823.1 | - | 4 | 289 | |||
| CB109881.1 | - | 4 | 233 | |||

C19orf33
Genomic Map