siRNA Details
| Gene Symbol: TBC1D3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123392.3 | NP_001116864.3 | 14 | 2078 | ||
| Gene Symbol: TBC1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123391.3 | NP_001116863.3 | 14 | 2076 | ||

TBC1D3H
Genomic Map