siRNA Details
| Gene Symbol: KRTAP4-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033187.1 | NP_149443.1 | 1 | 834 | ||
| GenBank mRNA | AJ406935.1 | - | 1 | 543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-3| Gene Symbol: KRTAP4-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033187.1 | NP_149443.1 | 1 | 834 | ||
| GenBank mRNA | AJ406935.1 | - | 1 | 543 | ||