siRNA Details
| Gene Symbol: ZNF541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277075.1 | NP_001264004.1 | 1 | 56 | ||
| XM_005259311.4 | XP_005259368.1 | 2 | 996 | |||
| XM_005259314.3 | XP_005259371.1 | 2 | 996 | |||
| XM_011527367.2 | XP_011525669.1 | 2 | 996 | |||
| XM_011527368.2 | XP_011525670.1 | 3 | 334 | |||
| XM_011527369.2 | XP_011525671.1 | 3 | 338 | |||
| XM_011527370.2 | XP_011525672.1 | 2 | 195 | |||
| XM_011527371.2 | XP_011525673.1 | 2 | 182 | |||
| XM_011527372.2 | XP_011525674.1 | 2 | 996 | |||
| XM_011527373.2 | XP_011525675.1 | 2 | 996 | |||
| XM_011527375.1 | XP_011525677.1 | 2 | 996 | |||
| XM_017027351.1 | XP_016882840.1 | 2 | 996 | |||

ZNF541
Genomic Map