siRNA Details
| Gene Symbol: KIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015074.3 | NP_055889.2 | 12 | 1309 | ||
| NM_183416.3 | NP_904325.2 | 12 | 1309 | |||
| GenBank mRNA | AB011163.2 | - | 13 | 1276 | ||
| AB017133.1 | - | 12 | 1159 | |||
| AB040881.2 | - | 12 | 1368 | |||
| AB088210.1 | - | 12 | 1075 | |||
| AB088211.1 | - | 12 | 1075 | |||
| AB088212.1 | - | 12 | 1075 | |||
| AB088213.1 | - | 12 | 1075 | |||
| AF257176.1 | - | 12 | 1210 | |||
| AY043362.1 | - | 11 | 1061 | |||
| AY139835.1 | - | 12 | 1210 | |||
| BC115395.2 | - | 11 | 1145 | 40067993 | ||

KIF1B
Genomic Map