siRNA Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207409.3 | NP_997292.2 | 2, 3 | 239 | ||
| NR_104467.1 | 2, 3 | 341 | ||||
| GenBank mRNA | CD513822.1 | - | 2, 3 | 368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLPSL2| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207409.3 | NP_997292.2 | 2, 3 | 239 | ||
| NR_104467.1 | 2, 3 | 341 | ||||
| GenBank mRNA | CD513822.1 | - | 2, 3 | 368 | ||