siRNA Details
| Gene Symbol: MUC5AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304359.1 | NP_001291288.1 | 1779 | |||
| GenBank mRNA | AF015521.1 | - | 15 | 1779 | ||
| AF043909.1 | - | 15 | 1770 | |||
| AJ298317.1 | - | 15 | 1770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC5AC| Gene Symbol: MUC5AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304359.1 | NP_001291288.1 | 1779 | |||
| GenBank mRNA | AF015521.1 | - | 15 | 1779 | ||
| AF043909.1 | - | 15 | 1770 | |||
| AJ298317.1 | - | 15 | 1770 | |||