siRNA Details
Gene Symbol: ETFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000126.3 | NP_000117.1 | 11 | 1030 | ||
NM_001127716.1 | NP_001121188.1 | 10 | 883 | |||
GenBank mRNA | AK292979.1 | - | 11 | 1030 | ||
AK300044.1 | - | 10 | 818 | |||
BC015526.1 | - | 11 | 974 | 3852346 | ||
BC029995.1 | - | 11 | 1019 | |||
BC034156.1 | - | 7 | 561 | |||
BC095457.1 | - | 11 | 955 | 30552923 | ||
BT009796.1 | - | 11 | 949 | |||
J04058.1 | - | 11 | 949 | |||
KU178046.1 | - | 11 | 949 | |||
KU178047.1 | - | 12 | 1082 | |||
KU178048.1 | - | 10 | 880 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145615, BY435583, AV289509, AK032830, AK028118, and others.
Panther Classification:
Gene Ontology Categories:
