siRNA Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 1544 | ||
| GenBank mRNA | AK058186.1 | - | 4 | 1155 | ||
| AK299804.1 | - | 1 | 753 | |||
| BC032356.1 | - | 2 | 1544 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM13| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 1544 | ||
| GenBank mRNA | AK058186.1 | - | 4 | 1155 | ||
| AK299804.1 | - | 1 | 753 | |||
| BC032356.1 | - | 2 | 1544 | |||