siRNA Details
| Gene Symbol: ECD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135752.1 | NP_001129224.1 | 2 | 301 | ||
| NM_001135753.1 | NP_001129225.1 | 2 | 301 | |||
| NM_007265.2 | NP_009196.1 | 2 | 301 | |||
| GenBank mRNA | AK022616.1 | - | 2 | 249 | ||
| AK225519.1 | - | 2 | 277 | |||
| AK315711.1 | - | 2 | 274 | |||
| BC000721.2 | - | 2 | 265 | 3504956 | ||
| CR457407.1 | - | 1 | 58 | |||
| D88208.1 | - | 2 | 265 | |||
| DA162516.1 | - | 2 | 277 | |||

ECD
Genomic Map