siRNA Details
| Gene Symbol: CCDC137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199287.2 | NP_954981.1 | 2, 3 | 315 | ||
| XM_017024573.1 | XP_016880062.1 | 2, 3 | 349 | |||
| GenBank mRNA | BC009369.2 | - | 2, 3 | 299 | 4110481 | |
| BM548065.1 | - | 2, 3 | 354 | |||
| FJ649645.1 | - | 2, 3 | 2005 | |||

CCDC137
Genomic Map