siRNA Details
| Gene Symbol: RPL27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000990.4 | NP_000981.1 | 4 | 589 | ||
| GenBank mRNA | AK311767.1 | - | 4 | 245 | ||
| BC005326.1 | - | 1 | 245 | 4052417 | ||
| BC020169.1 | - | 3 | 221 | |||
| BC066326.1 | - | 1 | 245 | 5311530 | ||
| BG687024.1 | - | 279 | ||||
| BQ058236.1 | - | 4 | 267 | |||
| U14968.1 | - | 4 | 239 | |||

RPL27A
Genomic Map